N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide 98%

BD154079-1G BD154079-250MG BD154079-100MG
BLDPBD154079-1GEA 569 EUR
BLDPBD154079-1G BLDPBD154079-250MG BLDPBD154079-100MG
N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide 98%
N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₀ClN₃OS
Molecular Weight: 267,73 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00113740
CAS-Nummer: 223580-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR