Thymolphthalein 96%

Synonyme: 2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton,3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid

BD154067-25G BD154067-5G BD154067-100G
BLDPBD154067-25GEA 11 EUR
BLDPBD154067-25G BLDPBD154067-5G BLDPBD154067-100G
Thymolphthalein 96%
Thymolphthalein

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₈H₃₀O₄
Molecular Weight: 430,54 g/mol
Lagertemperatur: Ambient
MDL: MFCD00005909
CAS-Nummer: 125-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR