Thiomorpholin-3-one 95%

BD153923-250MG BD153923-5G BD153923-1G
BLDPBD153923-250MGEA 21 EUR
BLDPBD153923-250MG BLDPBD153923-5G BLDPBD153923-1G
Thiomorpholin-3-one 95%
Thiomorpholin-3-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₄H₇NOS
Molecular Weight: 117,1700 g/mol
Lagertemperatur: Ambient
MDL: MFCD00152870
CAS-Nummer: 20196-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR