2-Allyl-4,6-dibenzoylresorcinol 98%

Synonyme: (5-Allyl-4,6-dihydroxy-1,3-phenylene)bis(phenylmethanone)

BD1538-100G BD1538-5G BD1538-25G BD1538-1G
BLDPBD1538-100GEA 899 EUR
BLDPBD1538-100G BLDPBD1538-5G BLDPBD1538-25G BLDPBD1538-1G
2-Allyl-4,6-dibenzoylresorcinol 98%
2-Allyl-4,6-dibenzoylresorcinol

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₃H₁₈O₄
Molecular Weight: 358,40 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD02094038
CAS-Nummer: 102593-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR