3,3'-Dimethylazobenzene 98%

Synonyme: m-Azotoluene,1,2-Di-m-tolyldiazene

BD15368-1G BD15368-250MG BD15368-5G BD15368-100MG
BLDPBD15368-1GEA 111 EUR
BLDPBD15368-1G BLDPBD15368-250MG BLDPBD15368-5G BLDPBD15368-100MG
3,3'-Dimethylazobenzene 98%
3,3'-Dimethylazobenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₄N₂
Molecular Weight: 210,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00048080
CAS-Nummer: 588-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR