Theacrine 97%

Synonyme: Temorine,1,3,7,9-Tetramethyluric acid,1,3,7,9-Tetramethyl-1H-purine-2,6,8(3H,7H,9H)-trione

BD153624-250MG BD153624-5G BD153624-1G BD153624-25G
BLDPBD153624-250MGEA 6 EUR
BLDPBD153624-250MG BLDPBD153624-5G BLDPBD153624-1G BLDPBD153624-25G
Theacrine 97%
Theacrine

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₂N₄O₃
Molecular Weight: 224,2 g/mol
Lagertemperatur: Ambient
MDL: MFCD00042781
CAS-Nummer: 2309-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR