Tetramethylammoniumhydrogensulfat Monohydrat 98%

Synonyme: Tetramethylammonium hydrogensulphate hydrate

BD153577-100G BD153577-25G BD153577-500G BD153577-5G
BLDPBD153577-100GEA 80 EUR
BLDPBD153577-100G BLDPBD153577-25G BLDPBD153577-500G BLDPBD153577-5G
Tetramethylammoniumhydrogensulfat Monohydrat 98%
Tetramethylammoniumhydrogensulfat Monohydrat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₄H₁₃NO₄S·1H₂O
Molecular Weight: 189,23 g/mol
Lagertemperatur: Ambient
MDL: MFCD00150356
CAS-Nummer: 103812-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR