Tetrakis(triphenylphosphin)platin(0) 98%

BD153562-1G BD153562-5G BD153562-25G
BLDPBD153562-1GEA 45 EUR
BLDPBD153562-1G BLDPBD153562-5G BLDPBD153562-25G
Tetrakis(triphenylphosphin)platin(0) 98%
Tetrakis(triphenylphosphin)platin(0)

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇₂H₆₀P₄Pt
Molecular Weight: 1244,24 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00010014
CAS-Nummer: 14221-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR