Tetrahydro-2H-thiopyran-4-ol 97%

BD153512-5G BD153512-1G BD153512-250MG BD153512-100MG BD153512-25G
BLDPBD153512-5GEA 238 EUR
BLDPBD153512-5G BLDPBD153512-1G BLDPBD153512-250MG BLDPBD153512-100MG BLDPBD153512-25G
Tetrahydro-2H-thiopyran-4-ol 97%
Tetrahydro-2H-thiopyran-4-ol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₁₀OS
Molecular Weight: 118,2 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD02942489
CAS-Nummer: 29683-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR