2-Isopropyl-5-methylhexan-1-ol 95+%

BLDPBD153490-1GEA 542 EUR
BLDPBD153490-1G BLDPBD153490-250MG BLDPBD153490-100MG
2-Isopropyl-5-methylhexan-1-ol 95+%
2-Isopropyl-5-methylhexan-1-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₂O
Molecular Weight: 158,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067466
CAS-Nummer: 2051-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR