Tetraethylethylenbisphosphonat 97%

Synonyme: Tetraethyl ethylenediphosphonate,Tetraethyl ethane-1,2-diylbis(phosphonate)

BD153337-1G BD153337-5G BD153337-25G
BLDPBD153337-1GEA 23 EUR
BLDPBD153337-1G BLDPBD153337-5G BLDPBD153337-25G
Tetraethylethylenbisphosphonat 97%
Tetraethylethylenbisphosphonat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₄O₆P₂
Molecular Weight: 302,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015133
CAS-Nummer: 995-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR