5,5'-sulphonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene) 95+%

BD153222-5G BD153222-1G BD153222-250MG BD153222-100MG
BLDPBD153222-5GEA 1031 EUR
BLDPBD153222-5G BLDPBD153222-1G BLDPBD153222-250MG BLDPBD153222-100MG
5,5'-sulphonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene) 95+%
5,5'-sulphonylbis(1,3-dibromo-2-(2,3-dibromopropoxy)benzene)

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₁₄Br₈O₄S
Molecular Weight: 965,59 g/mol
Lagertemperatur: Ambient
MDL: MFCD00329083
CAS-Nummer: 42757-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR