2-tert-butyl-1,4-phenylene diacetate 98%

BD153154-5G BD153154-1G BD153154-25G
BLDPBD153154-5GEA 96 EUR
BLDPBD153154-5G BLDPBD153154-1G BLDPBD153154-25G
2-tert-butyl-1,4-phenylene diacetate 98%
2-tert-butyl-1,4-phenylene diacetate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₈O₄
Molecular Weight: 250,29 g/mol
Lagertemperatur: Ambient
MDL: MFCD00078251
CAS-Nummer: 7507-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR