Pivalamidine hydrochloride 97%

BLDPBD153132-1GEA 14 EUR
BLDPBD153132-1G BLDPBD153132-250MG BLDPBD153132-100G BLDPBD153132-500G BLDPBD153132-25G BLDPBD153132-5G BLDPBD153132-10G
Pivalamidine hydrochloride 97%
Pivalamidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₂N₂
Molecular Weight: 136,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051988
CAS-Nummer: 18202-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR