tert-Butyl-4-chloro-3-oxobutanoate 95%

BD153121-100MG BD153121-250MG BD153121-5G BD153121-1G
BLDPBD153121-100MGEA 70 EUR
BLDPBD153121-100MG BLDPBD153121-250MG BLDPBD153121-5G BLDPBD153121-1G
tert-Butyl-4-chloro-3-oxobutanoate 95%
tert-Butyl-4-chloro-3-oxobutanoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₁₃ClO₃
Molecular Weight: 192,64 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00052188
CAS-Nummer: 74530-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR