tert-Butyl 2-bromobenzoate 97%

BD153105-25G BD153105-100G BD153105-5G BD153105-1G
BLDPBD153105-25GEA 312 EUR
BLDPBD153105-25G BLDPBD153105-100G BLDPBD153105-5G BLDPBD153105-1G
tert-Butyl 2-bromobenzoate 97%
tert-Butyl 2-bromobenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD00027950
CAS-Nummer: 55666-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR