tert-Butyl(4-iodobutoxy)dimethylsilane 95%

BD153100-1G BD153100-5G BD153100-250MG
BLDPBD153100-1GEA 46 EUR
BLDPBD153100-1G BLDPBD153100-5G BLDPBD153100-250MG
tert-Butyl(4-iodobutoxy)dimethylsilane 95%
tert-Butyl(4-iodobutoxy)dimethylsilane

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₂₃IOSi
Molecular Weight: 314,279 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01863695
CAS-Nummer: 92511-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR