tert-Butyl(4-fluorophenoxy)dimethylsilane 95+%

BD153099-1G BD153099-5G BD153099-250MG
BLDPBD153099-1GEA 77 EUR
BLDPBD153099-1G BLDPBD153099-5G BLDPBD153099-250MG
tert-Butyl(4-fluorophenoxy)dimethylsilane 95+%
tert-Butyl(4-fluorophenoxy)dimethylsilane

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₉FOSi
Molecular Weight: 226,36 g/mol
Lagertemperatur: Ambient
MDL: MFCD08704818
CAS-Nummer: 113984-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR