4-(Benzyloxy)-2-hydroxybenzaldehyd 97%

BD15308-250MG BD15308-100MG BD15308-5G BD15308-25G BD15308-1G
BLDPBD15308-250MGEA 9 EUR
BLDPBD15308-250MG BLDPBD15308-100MG BLDPBD15308-5G BLDPBD15308-25G BLDPBD15308-1G
4-(Benzyloxy)-2-hydroxybenzaldehyd 97%
4-(Benzyloxy)-2-hydroxybenzaldehyd

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD01075698
CAS-Nummer: 52085-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR