tert-Butylcyanacetat 98%

Synonyme: Cyanessigsäure-tert-butylester

BD153001-500G BD153001-100G BD153001-25G BD153001-10G BD153001-5G
BLDPBD153001-500GEA 124 EUR
BLDPBD153001-500G BLDPBD153001-100G BLDPBD153001-25G BLDPBD153001-10G BLDPBD153001-5G
tert-Butylcyanacetat 98%
tert-Butylcyanacetat

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00001938
CAS-Nummer: 1116-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR