2-Benzylbenzoic acid 98%

Synonyme: α-Phenyl-o-toluic acid,2-Carboxydiphenylmethane

BD15299-5G BD15299-100G BD15299-25G BD15299-10G
BLDPBD15299-5GEA 29 EUR
BLDPBD15299-5G BLDPBD15299-100G BLDPBD15299-25G BLDPBD15299-10G
2-Benzylbenzoic acid 98%
2-Benzylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₂O₂
Molecular Weight: 212,244 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002484
CAS-Nummer: 612-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR