tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate 95%

BD152982-1G BD152982-250MG BD152982-100MG BD152982-5G
BLDPBD152982-1GEA 190 EUR
BLDPBD152982-1G BLDPBD152982-250MG BLDPBD152982-100MG BLDPBD152982-5G
tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate 95%
tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₄H₁₉NO₃
Molecular Weight: 249,30 g/mol
Lagertemperatur: Freezer
MDL: MFCD08275030
CAS-Nummer: 676255-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR