Methyl-4-phenylbenzoat 95%

Synonyme: p-Phenylbenzoic acid-OMe

BLDPBD15297-500GEA 985 EUR
BLDPBD15297-500G BLDPBD15297-5G BLDPBD15297-1G BLDPBD15297-100G BLDPBD15297-25G
Methyl-4-phenylbenzoat 95%
Methyl-4-phenylbenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017200
CAS-Nummer: 720-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR