tert-Butyl 4-bromo-2-methylbenzoate 98%

BD152918-100MG BD152918-25G BD152918-250MG BD152918-5G BD152918-1G
BLDPBD152918-100MGEA 42 EUR
BLDPBD152918-100MG BLDPBD152918-25G BLDPBD152918-250MG BLDPBD152918-5G BLDPBD152918-1G
tert-Butyl 4-bromo-2-methylbenzoate 98%
tert-Butyl 4-bromo-2-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₅BrO₂
Molecular Weight: 271,15 g/mol
Lagertemperatur: Ambient
MDL: MFCD08703427
CAS-Nummer: 445003-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR