3'-Methoxybiphenyl-3-carbaldehyde 98%

BLDPBD15290-250MGEA 69 EUR
BLDPBD15290-250MG BLDPBD15290-100MG
3'-Methoxybiphenyl-3-carbaldehyde 98%
3'-Methoxybiphenyl-3-carbaldehyde
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD03424626
CAS-Nummer: 126485-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR