3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%

BLDPBD152830-10GEA 494 EUR
BLDPBD152830-10G BLDPBD152830-1G BLDPBD152830-250MG BLDPBD152830-5G BLDPBD152830-100MG
3-Amino-N-(tert-butoxycarbonyl)benzylamine 97%
3-Amino-N-(tert-butoxycarbonyl)benzylamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04974010
CAS-Nummer: 147291-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR