3-Acetylbiphenyl 97%

Synonyme: 1-Biphenyl-3-yl-ethanone,1-([1,1'-Biphenyl]-3-yl)ethanone

BD15280-1G BD15280-5G BD15280-250MG
BLDPBD15280-1GEA 45 EUR
BLDPBD15280-1G BLDPBD15280-5G BLDPBD15280-250MG
3-Acetylbiphenyl 97%
3-Acetylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₂O
Molecular Weight: 196,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD00048220
CAS-Nummer: 3112-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR