tert-Butyl-4-brom-2-fluorbenzoat 98%
|
|
BLDPBD152786-100GEA
502
EUR
BLDPBD152786-100G
BLDPBD152786-10G
BLDPBD152786-25G
BLDPBD152786-5G
BLDPBD152786-1G
tert-Butyl-4-brom-2-fluorbenzoat 98%
tert-Butyl-4-brom-2-fluorbenzoat
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₁H₁₂BrFO₂ Molecular Weight: 275,1100 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08703426 CAS-Nummer: 889858-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...