Ethyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 97%

Synonyme: N-(tert-Butoxycarbonyl)glycine ethyl ester,ethyl 2-((tert-Butoxycarbonyl)amino)acetate,Boc-Gly-OEt,N-Boc-glycine ethyl ester

BD152685-5G BD152685-10G BD152685-100G BD152685-500G BD152685-25G
BLDPBD152685-5GEA 6 EUR
BLDPBD152685-5G BLDPBD152685-10G BLDPBD152685-100G BLDPBD152685-500G BLDPBD152685-25G
Ethyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 97%
ethyl-2-((tert-butoxycarbonyl)amino)acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₇NO₄
Molecular Weight: 203,2 g/mol
Lagertemperatur: Ambient
MDL: MFCD05663720
CAS-Nummer: 14719-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR