Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) 98%

Synonyme: Cu(TMHD)2,Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate),Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)

BD1525-1G BD1525-5G BD1525-10G
BLDPBD1525-1GEA 16 EUR
BLDPBD1525-1G BLDPBD1525-5G BLDPBD1525-10G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')kupfer(II)

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₂H₃₈CuO₄
Molecular Weight: 430,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD00058920
CAS-Nummer: 14040-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR