Taurolidine 97%

Synonyme: 4,4'-Methylenebis(1,2,4-thiadiazinane 1,1-dioxide)

BD152420-5G BD152420-25G BD152420-1G
BLDPBD152420-5GEA 29 EUR
BLDPBD152420-5G BLDPBD152420-25G BLDPBD152420-1G
Taurolidine 97%
Taurolidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₁₆N₄O₄S₂
Molecular Weight: 284,4 g/mol
Lagertemperatur: Ambient
MDL: MFCD00865076
CAS-Nummer: 19388-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR