Sulindac 98%

Synonyme: (Z)-2-(5-Fluoro-2-methyl-1-(4-(methylsulphinyl)benzylidene)-1H-inden-3-yl)acetic acid

BD152239-5G BD152239-25G
BLDPBD152239-5GEA 17 EUR
BLDPBD152239-5G BLDPBD152239-25G
Sulindac 98%
Sulindac

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₀H₁₇FO₃S
Molecular Weight: 356,411 g/mol
Lagertemperatur: Freezer
MDL: MFCD00599589
CAS-Nummer: 38194-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR