Sulphachloropyridazine sodium 98%

BLDPBD152137-500GEA 132 EUR
BLDPBD152137-500G BLDPBD152137-100G BLDPBD152137-25G BLDPBD152137-1000G
Sulphachloropyridazine sodium 98%
Sulphachloropyridazine sodium

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₈ClN₄NaO₂S
Molecular Weight: 306,70 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04117866
CAS-Nummer: 23282-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR