Sucralfate 30-38%

Synonyme: Hexadeca-μ-hydroxytetracosahydroxy[μ8-[[1,3,4,6-tetra-O-sulpho-β-D-fructofuranosyl α-D-glucopyranoside tetrakis(sulphato-κO′)](8-)]]hexadecaaluminum

BD152091-25G BD152091-500G BD152091-100G BD152091-1000G
BLDPBD152091-25GEA 6 EUR
BLDPBD152091-25G BLDPBD152091-500G BLDPBD152091-100G BLDPBD152091-1000G
Sucralfate 30-38%
Sucralfate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₅₄Al₁₆O₇₅S₈
Molecular Weight: 2086,7368 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00145339
CAS-Nummer: 54182-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR