Succinamid 98%

BD152058-100G BD152058-5G BD152058-25G
BLDPBD152058-100GEA 95 EUR
BLDPBD152058-100G BLDPBD152058-5G BLDPBD152058-25G
Succinamid 98%
Succinamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₄H₈N₂O₂
Molecular Weight: 116,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008042
CAS-Nummer: 110-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR