Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate 98%

Synonyme: 2,2'-((5-Carboxy-4-(carboxymethyl)-3-cyanothiophen-2-yl)azanediyl)diacetic acid, distrontium salt

BD151999-10G BD151999-500G BD151999-100G BD151999-5G BD151999-25G
BLDPBD151999-10GEA 29 EUR
BLDPBD151999-10G BLDPBD151999-500G BLDPBD151999-100G BLDPBD151999-5G BLDPBD151999-25G
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate 98%
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₂H₆N₂O₈SSr₂
Molecular Weight: 513,49 g/mol
Lagertemperatur: Freezer
MDL: MFCD09038742
CAS-Nummer: 135459-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR