2',3'-Didehydro-3'-deoxythymidine 95%

BLDPBD151900-10GEA 45 EUR
BLDPBD151900-10G BLDPBD151900-5G BLDPBD151900-25G BLDPBD151900-1G BLDPBD151900-100G
2',3'-Didehydro-3'-deoxythymidine 95%
2',3'-Didehydro-3'-deoxythymidine

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₀H₁₂N₂O₄
Molecular Weight: 224,22 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00132921
CAS-Nummer: 3056-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR