SQ 22536 95%

Synonyme: 9-(Tetrahydro-2-furyl)adenine,9-(Tetrahydro-2-furanyl)-9H-purin-6-amine,9-(Tetrahydrofuran-2-yl)-9H-purin-6-amine

BD151862-250MG BD151862-1G BD151862-50MG BD151862-100MG
BLDPBD151862-250MGEA 291 EUR
BLDPBD151862-250MG BLDPBD151862-1G BLDPBD151862-50MG BLDPBD151862-100MG
SQ 22536 95%
SQ 22536

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₁₁N₅O
Molecular Weight: 205,221 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00210216
CAS-Nummer: 17318-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR