Spiro[cyclopropane-1,1'-isoindolin]-3'-one 98%

BD151842-100MG BD151842-250MG BD151842-50MG
BLDPBD151842-100MGEA 576 EUR
BLDPBD151842-100MG BLDPBD151842-250MG BLDPBD151842-50MG
Spiro[cyclopropane-1,1'-isoindolin]-3'-one 98%
Spiro[cyclopropane-1,1'-isoindolin]-3'-one

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₉NO
Molecular Weight: 159,18 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08234398
CAS-Nummer: 604799-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR