4-Benzoylbenzoesäure 98%

Synonyme: Benzophenon-4-carbonsäure

BD15181-10G BD15181-1G BD15181-250MG BD15181-5G BD15181-25G BD15181-100G
BLDPBD15181-10GEA 88 EUR
BLDPBD15181-10G BLDPBD15181-1G BLDPBD15181-250MG BLDPBD15181-5G BLDPBD15181-25G BLDPBD15181-100G
4-Benzoylbenzoesäure 98%
4-Benzoylbenzoesäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₀O₃
Molecular Weight: 226,23 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002560
CAS-Nummer: 611-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR