9-Fluorencarbonsäure 95%

Synonyme: 9H-Fluorene-9-carboxylic acid

BD15180-100G BD15180-5G BD15180-25G BD15180-1G BD15180-250MG BD15180-500G BD15180-10G
BLDPBD15180-100GEA 134 EUR
BLDPBD15180-100G BLDPBD15180-5G BLDPBD15180-25G BLDPBD15180-1G BLDPBD15180-250MG BLDPBD15180-500G BLDPBD15180-10G
9-Fluorencarbonsäure 95%
9-Fluorencarbonsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₀O₂
Molecular Weight: 210,23 g/mol
Lagertemperatur: Ambient
MDL: MFCD00001136
CAS-Nummer: 1989-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR