Sparfloxacin 99+%

Synonyme: 5-Amino-1-cyclopropyl-7-((3S,5R)-3,5-dimethylpiperazin-1-yl)-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

BD151779-25G BD151779-1G BD151779-100G BD151779-5G
BLDPBD151779-25GEA 116 EUR
BLDPBD151779-25G BLDPBD151779-1G BLDPBD151779-100G BLDPBD151779-5G
Sparfloxacin 99+%
Sparfloxacin

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₉H₂₂F₂N₄O₃
Molecular Weight: 392,407 g/mol
Lagertemperatur: Ambient
MDL: MFCD00869619
CAS-Nummer: 110871-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR