Sorbitantristearat 98+%

Synonyme: Sorbitantrioctadecanoat

BD151760-500G BD151760-100G BD151760-25G BD151760-1000G
BLDPBD151760-500GEA 15 EUR
BLDPBD151760-500G BLDPBD151760-100G BLDPBD151760-25G BLDPBD151760-1000G
Sorbitantristearat 98+%
Sorbitantristearat

Storage condition: Sealed in dry, Room Temperature

Formel: C₆₀H₁₁₄O₈
Molecular Weight: 963,56 g/mol
Lagertemperatur: Ambient
MDL: MFCD00151165
CAS-Nummer: 26658-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR