Calciumdi[(2E,4E)-2,4-hexadienoat] 97%

Synonyme: Calcium Sorbate,Sorbic acid calcium salt,Calcium (2E,4E)-hexa-2,4-dienoate

BD151752-25G
BLDPBD151752-25GEA 7 EUR
BLDPBD151752-25G
Calciumdi[(2E,4E)-2,4-hexadienoat] 97%
Calcium(2E,4E)hexa-2,4-dienoate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₄CaO₄
Molecular Weight: 262,3200 g/mol
Lagertemperatur: Ambient
MDL: MFCD00043805
CAS-Nummer: 7492-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR