Diisobutyl perylenedicarboxylate 95%

Synonyme: Diisobutyl perylene-3,9-dicarboxylatePerylenedicarboxylic acid Diisobutyl ester

BLDPBD151711-5GEA 6 EUR
BLDPBD151711-5G BLDPBD151711-500G BLDPBD151711-10G BLDPBD151711-25G BLDPBD151711-100G
Diisobutyl perylenedicarboxylate 95%
Diisobutyl perylenedicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₀H₂₈O₄
Molecular Weight: 452,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191681
CAS-Nummer: 2744-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR