Sodium trimethylsilanolate 95%

Synonyme: Silanol, 1,1,1-trimethyl-, sodium salt (1:1),sodium trimethyl(oxido)silane,Silanol, trimethyl-, sodium salt

BD151674-250MG
BLDPBD151674-250MGEA 8 EUR
BLDPBD151674-250MG
Sodium trimethylsilanolate 95%
Sodium trimethylsilanolate
Formel: C₃H₉NaOSi
Molecular Weight: 112,179 g/mol
MDL: MFCD00013926
CAS-Nummer: 18027-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR