Sodium pivalate hydrate 95%

Synonyme: Trimethylacetic acid sodium salt hydrate,Sodium Trimethylacetate xHydrate,Sodium trimethylacetate hydrate,Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate

BD151673-25G BD151673-5G BD151673-10G BD151673-500G BD151673-100G
BLDPBD151673-25GEA 25 EUR
BLDPBD151673-25G BLDPBD151673-5G BLDPBD151673-10G BLDPBD151673-500G BLDPBD151673-100G
Sodium pivalate hydrate 95%
Sodium pivalate hydrate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₁NaO₃
Molecular Weight: 142,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD00150782
CAS-Nummer: 143174-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR