Sodiumthiophosphatedodecahydrate 97%

BLDPBD151660-500GEA 457 EUR
BLDPBD151660-500G BLDPBD151660-5G BLDPBD151660-1G BLDPBD151660-25G BLDPBD151660-100G
Sodiumthiophosphatedodecahydrate 97%
Sodiumthiophosphatedodecahydrate

Storage condition: Inert atmosphere, Room Temperature

Formel: H₂₄Na₃O₁₅PS
Molecular Weight: 396,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150620
CAS-Nummer: 51674-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR