Sodium stearyl fumarate 95%

Synonyme: Fumaric Acid Stearyl Ester Sodium Salt,Fumaric Acid Octadecyl Ester Sodium Salt,Sodium Octadecyl Fumarate,Sodium 2-octadecylfumarate

BD151619-25G BD151619-10G BD151619-5G
BLDPBD151619-25GEA 22 EUR
BLDPBD151619-25G BLDPBD151619-10G BLDPBD151619-5G
Sodium stearyl fumarate 95%
Sodium stearyl fumarate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₂H₄₀O₄
Molecular Weight: 390,54 g/mol
Lagertemperatur: Ambient
MDL: MFCD00236640
CAS-Nummer: 4070-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR