Natriumoleat 97%

Synonyme: Ölsäure Natriumsalzcis-9-Octadecensäuremethylester Natriumsalz

BLDPBD151556-500GEA 25 EUR
BLDPBD151556-500G BLDPBD151556-0.5G BLDPBD151556-1000G BLDPBD151556-100G BLDPBD151556-25G BLDPBD151556-10G
Natriumoleat 97%
Natriumoleat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₈H₃₃NaO₂
Molecular Weight: 304,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004438
CAS-Nummer: 143-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR